siRNA Details
| Gene Symbol: Rnf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027258.1 | NP_081534.1 | 3 | 513 | ||
| GenBank mRNA | BC053070.1 | - | 3 | 292 | ||
| CF539215.1 | - | 2 | 155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf157| Gene Symbol: Rnf157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027258.1 | NP_081534.1 | 3 | 513 | ||
| GenBank mRNA | BC053070.1 | - | 3 | 292 | ||
| CF539215.1 | - | 2 | 155 | |||