siRNA Details
| Gene Symbol: Qrich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033267.2 | NP_001028439.1 | 10 | 1340 | ||
| GenBank mRNA | AK131945.1 | - | 10 | 1343 | ||
| BC150976.1 | - | 10 | 1193 | 9087889 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qrich2| Gene Symbol: Qrich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033267.2 | NP_001028439.1 | 10 | 1340 | ||
| GenBank mRNA | AK131945.1 | - | 10 | 1343 | ||
| BC150976.1 | - | 10 | 1193 | 9087889 | ||