siRNA Details
| Gene Symbol: Plekhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177503.1 | NP_001170974.1 | 11 | 1422 | ||
| GenBank mRNA | AK028383.1 | - | 11 | 1372 | ||
| BC137602.1 | - | 11 | 1205 | 8860621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhd1| Gene Symbol: Plekhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177503.1 | NP_001170974.1 | 11 | 1422 | ||
| GenBank mRNA | AK028383.1 | - | 11 | 1372 | ||
| BC137602.1 | - | 11 | 1205 | 8860621 | ||