siRNA Details
| Gene Symbol: Fam120a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033268.2 | NP_001028440.2 | 18 | 4123 | ||
| GenBank mRNA | AK035074.1 | - | 8 | 2037 | ||
| AK172909.1 | - | 18 | 4312 | |||
| BC010304.1 | - | 14 | 3194 | |||
| BC033412.1 | - | 10 | 2453 | |||
| BC042582.1 | - | 17 | 3641 | |||

Fam120a
Genomic Map