siRNA Details
| Gene Symbol: Akap11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164503.1 | NP_001157975.1 | 12 | 7688 | ||
| GenBank mRNA | AK078114.1 | - | 1 | 1122 | ||
| AK129178.1 | - | 2 | 4152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap11| Gene Symbol: Akap11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164503.1 | NP_001157975.1 | 12 | 7688 | ||
| GenBank mRNA | AK078114.1 | - | 1 | 1122 | ||
| AK129178.1 | - | 2 | 4152 | |||