siRNA Details
| Gene Symbol: Pex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017234.1 | NP_058930.1 | 3 | 587 | ||
| XM_006232157.2 | XP_006232219.1 | 2 | 1676 | |||
| XM_006232158.1 | XP_006232220.1 | 3 | 439 | |||
| XM_006232159.1 | XP_006232221.1 | 3 | 695 | |||
| XM_006232160.2 | XP_006232222.1 | 3 | 1776 | |||
| XM_006232161.1 | XP_006232223.1 | 3 | 705 | |||
| XM_008760865.1 | XP_008759087.1 | 3 | 456 | |||
| XM_008760866.1 | XP_008759088.1 | 2 | 352 | |||
| GenBank mRNA | BC063169.1 | - | 3 | 437 | 6886755 | |
| D30616.1 | - | 3 | 554 | |||
| D30617.1 | - | 3 | 587 | |||
| X57988.1 | - | 2 | 363 | |||

Pex2
Genomic Map