siRNA Details
| Gene Symbol: Trio | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081302.1 | NP_001074771.1 | 57 | 11395 | ||
| GenBank mRNA | BC035955.1 | - | 8 | 3027 | ||
| BC051169.1 | - | 16 | 4862 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_006232091, XM_003749226, XM_006232090.

Trio
Genomic Map