siRNA Details
| Gene Symbol: Hsd17b7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017235.3 | NP_058931.1 | 2, 3 | 302 | ||
| XM_006250215.2 | XP_006250277.1 | 2, 3 | 325 | |||
| XM_006250216.2 | XP_006250278.1 | 2, 3 | 325 | |||
| XM_006250217.2 | XP_006250279.1 | 2, 3 | 325 | |||
| GenBank mRNA | FM073997.1 | - | 2, 3 | 295 | ||
| FQ113345.1 | - | 2, 3 | 302 | |||
| U44803.1 | - | 2, 3 | 247 | |||

Hsd17b7
Genomic Map