siRNA Details
| Gene Symbol: Mroh2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166066.1 | NP_001159538.1 | 30 | 3535 | ||
| GenBank mRNA | KF778378.1 | - | 29 | 3115 | ||
| KF778379.1 | - | 9 | 1135 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mroh2b| Gene Symbol: Mroh2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166066.1 | NP_001159538.1 | 30 | 3535 | ||
| GenBank mRNA | KF778378.1 | - | 29 | 3115 | ||
| KF778379.1 | - | 9 | 1135 | |||