siRNA Details
| Gene Symbol: Gxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033275.4 | NP_001028447.1 | 8 | 1728 | ||
| GenBank mRNA | AK147387.1 | - | 8 | 1728 | ||
| AK163581.1 | - | 1 | 37 | |||
| BC141302.1 | - | 7 | 1733 | 9055914 | ||
| BC171972.1 | - | 7 | 1733 | 9054258 | ||

Gxylt1
Genomic Map