siRNA Details
| Gene Symbol: Gxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033275.4 | NP_001028447.1 | 8 | 5734 | ||
| GenBank mRNA | AK147387.1 | - | 8 | 5734 | ||
| AK161787.1 | - | 1 | 369 | |||
| BC024541.1 | - | 1 | 50 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gxylt1| Gene Symbol: Gxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033275.4 | NP_001028447.1 | 8 | 5734 | ||
| GenBank mRNA | AK147387.1 | - | 8 | 5734 | ||
| AK161787.1 | - | 1 | 369 | |||
| BC024541.1 | - | 1 | 50 | |||