siRNA Details
| Gene Symbol: Spryd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033277.3 | NP_001028449.2 | 5 | 544 | ||
| GenBank mRNA | AK146689.1 | - | 5 | 547 | ||
| BC008150.1 | - | 1 | 55 | |||
| BC048820.1 | - | 4 | 773 | |||
| BU057890.1 | - | 5 | 479 | |||
| CX201967.1 | - | 1 | 43 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001191790.

Spryd3
Genomic Map