siRNA Details
| Gene Symbol: Vipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017238.1 | NP_058934.1 | 2 | 222 | ||
| GenBank mRNA | U09631.1 | - | 2 | 222 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vipr2| Gene Symbol: Vipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017238.1 | NP_058934.1 | 2 | 222 | ||
| GenBank mRNA | U09631.1 | - | 2 | 222 | ||
Set Membership: |
|