siRNA Details
| Gene Symbol: Golgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030035.1 | NP_084311.1 | 22 | 10990 | ||
| GenBank mRNA | AK081762.1 | - | 3 | 1393 | ||
| AK220188.1 | - | 10 | 6845 | |||
| BC040461.1 | - | 10 | 4310 | |||
| BC042759.1 | - | 9 | 2325 | |||
| BC055037.1 | - | 1 | 1896 | |||
| BC060254.1 | - | 9 | 3412 | |||

Golgb1
Genomic Map