siRNA Details
| Gene Symbol: Dzip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110017.1 | NP_001103487.1 | 7 | 865 | ||
| NM_027341.2 | NP_081617.1 | 7 | 865 | |||
| GenBank mRNA | AK039172.1 | - | 7 | 843 | ||
| BC052893.1 | - | 7 | 673 | |||
| BC117953.1 | - | 7 | 688 | 40098141 | ||
| BC117954.1 | - | 7 | 694 | 40098143 | ||

Dzip3
Genomic Map