siRNA Details
| Gene Symbol: Dzip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110017.1 | NP_001103487.1 | 32 | 5469 | ||
| NM_027341.2 | NP_081617.1 | 31 | 4851 | |||
| GenBank mRNA | AK032456.1 | - | 1 | 3346 | ||
| AK083078.1 | - | 929 | ||||
| AK122344.1 | - | 7 | 5537 | |||
| AK160392.1 | - | 1 | 916 | |||
| AK163763.1 | - | 1 | 2741 | |||

Dzip3
Genomic Map