siRNA Details
| Gene Symbol: Scaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178923.3 | NP_849254.2 | 20 | 3895 | ||
| GenBank mRNA | AK045266.1 | - | 20 | 3883 | ||
| AK173120.1 | - | 20 | 3895 | |||
| BC023734.1 | - | 17 | 3293 | |||
| BC024860.1 | - | 2 | 1023 | |||
| BC053096.1 | - | 20 | 3712 | 6401143 | ||
| BC057592.1 | - | 20 | 3895 | 6417362 | ||

Scaf4
Genomic Map