siRNA Details
| Gene Symbol: D17Wsu92e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033279.3 | NP_001028451.2 | 5 | 1687 | ||
| NM_001044719.2 | NP_001038184.1 | 5 | 1462 | |||
| NM_001271511.1 | NP_001258440.1 | 4 | 1358 | |||
| GenBank mRNA | CF536829.1 | - | 2 | 715 | ||

D17Wsu92e
Genomic Map