siRNA Details
| Gene Symbol: Zfp81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207541.1 | NP_997424.1 | 4 | 1483 | ||
| GenBank mRNA | AK079439.1 | - | 1, 2 | 642 | ||
| AK135853.1 | - | 4 | 1484 | |||
| BB143786.2 | - | 1 | 100 | |||
| BC026640.1 | - | 1 | 906 | |||
| BC117831.1 | - | 4 | 1433 | 40092935 | ||
| BC117832.1 | - | 4 | 1435 | 40092936 | ||

Zfp81
Genomic Map