siRNA Details
| Gene Symbol: Ankrd29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190371.1 | NP_001177300.1 | 10 | 2523 | ||
| GenBank mRNA | AK162301.1 | - | 5 | 1954 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd29| Gene Symbol: Ankrd29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190371.1 | NP_001177300.1 | 10 | 2523 | ||
| GenBank mRNA | AK162301.1 | - | 5 | 1954 | ||