siRNA Details
Gene Symbol: Prdm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033281.3 | NP_001028453.1 | 5 | 1367 | ||
GenBank mRNA | AK133910.1 | - | 5 | 1368 | ||
AK135836.1 | - | 2 | 241 | |||
AK135840.1 | - | 2 | 241 | |||
BC139295.1 | - | 5 | 1397 | 8862317 | ||
DQ340564.1 | - | 5 | 943 | |||
DQ340565.1 | - | 5 | 862 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
