siRNA Details
| Gene Symbol: Cep76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081073.1 | NP_001074542.1 | 12 | 2520 | ||
| GenBank mRNA | AK035879.1 | - | 11 | 2300 | ||
| AK078034.1 | - | 2 | 664 | |||
| AK165893.1 | - | 2 | 558 | |||
| BC119216.1 | - | 12 | 2401 | 8733965 | ||
| BC119218.1 | - | 12 | 2401 | 8733967 | ||

Cep76
Genomic Map