siRNA Details
| Gene Symbol: Cep76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081073.1 | NP_001074542.1 | 5 | 784 | ||
| GenBank mRNA | AK035879.1 | - | 4 | 564 | ||
| BC119216.1 | - | 5 | 665 | 8733965 | ||
| BC119218.1 | - | 5 | 665 | 8733967 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep76| Gene Symbol: Cep76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081073.1 | NP_001074542.1 | 5 | 784 | ||
| GenBank mRNA | AK035879.1 | - | 4 | 564 | ||
| BC119216.1 | - | 5 | 665 | 8733965 | ||
| BC119218.1 | - | 5 | 665 | 8733967 | ||