siRNA Details
| Gene Symbol: Cep76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081073.1 | NP_001074542.1 | 12 | 2824 | ||
| GenBank mRNA | AK035879.1 | - | 11 | 2604 | ||
| AK078034.1 | - | 2 | 968 | |||
| AK165893.1 | - | 2 | 862 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep76| Gene Symbol: Cep76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081073.1 | NP_001074542.1 | 12 | 2824 | ||
| GenBank mRNA | AK035879.1 | - | 11 | 2604 | ||
| AK078034.1 | - | 2 | 968 | |||
| AK165893.1 | - | 2 | 862 | |||