siRNA Details
| Gene Symbol: Rnf165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164504.1 | NP_001157976.1 | 8 | 5374 | ||
| GenBank mRNA | AK090360.1 | - | 1 | 965 | ||
| BC066016.1 | - | 1 | 1164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf165| Gene Symbol: Rnf165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164504.1 | NP_001157976.1 | 8 | 5374 | ||
| GenBank mRNA | AK090360.1 | - | 1 | 965 | ||
| BC066016.1 | - | 1 | 1164 | |||