siRNA Details
| Gene Symbol: Map3k19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011737.1 | NP_035867.1 | 7 | 2656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k19| Gene Symbol: Map3k19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011737.1 | NP_035867.1 | 7 | 2656 | ||