siRNA Details
| Gene Symbol: Zranb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285945.1 | NP_001272874.1 | 21 | 3679 | ||
| NM_027678.3 | NP_081954.1 | 21 | 3818 | |||
| GenBank mRNA | AK050039.1 | - | 17 | 3099 | ||
| AK083641.1 | - | 21 | 3797 | |||
| BC066035.1 | - | 21 | 3755 | 6837116 | ||

Zranb3
Genomic Map