siRNA Details
| Gene Symbol: BC026585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033284.1 | NP_001028456.1 | 3 | 229 | ||
| GenBank mRNA | AK143913.1 | - | 3 | 229 | ||
| AK151310.1 | - | 3 | 237 | |||
| BC026585.1 | - | 3 | 231 | |||
| BC045198.1 | - | 3 | 226 | 5065108 | ||

BC026585
Genomic Map