siRNA Details
Gene Symbol: Sbspon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033288.3 | NP_001028460.1 | 5 | 1752 | ||
GenBank mRNA | AK137144.1 | - | 5 | 1754 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sbspon | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033288.3 | NP_001028460.1 | 5 | 1752 | ||
GenBank mRNA | AK137144.1 | - | 5 | 1754 |