siRNA Details
| Gene Symbol: Sbspon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033288.3 | NP_001028460.1 | 5 | 1005 | ||
| GenBank mRNA | AK137144.1 | - | 5 | 1006 | ||
| AK143251.1 | - | 5 | 979 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbspon| Gene Symbol: Sbspon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033288.3 | NP_001028460.1 | 5 | 1005 | ||
| GenBank mRNA | AK137144.1 | - | 5 | 1006 | ||
| AK143251.1 | - | 5 | 979 | |||