siRNA Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011753.3 | NP_035883.2 | 10 | 10339 | ||
| GenBank mRNA | AK133779.1 | - | 1 | 1052 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp26| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011753.3 | NP_035883.2 | 10 | 10339 | ||
| GenBank mRNA | AK133779.1 | - | 1 | 1052 | ||