siRNA Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011753.3 | NP_035883.2 | 10 | 9198 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp26| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011753.3 | NP_035883.2 | 10 | 9198 | ||