siRNA Details
| Gene Symbol: Usp40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033291.2 | NP_001028463.1 | 7 | 921 | ||
| NM_001198573.1 | NP_001185502.1 | 7 | 921 | |||
| GenBank mRNA | AK050239.1 | - | 7 | 918 | ||
| AK132295.1 | - | 7 | 921 | |||
| AK166303.1 | - | 7 | 898 | |||
| BC030843.1 | - | 7 | 870 | |||
| BC089376.1 | - | 4 | 423 | |||
| BC117957.1 | - | 8 | 1036 | 40098174 | ||
| BC117958.1 | - | 7 | 904 | 40098175 | ||

Usp40
Genomic Map