siRNA Details
| Gene Symbol: Espnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033292.3 | NP_001028464.1 | 9 | 3029 | ||
| GenBank mRNA | AK134448.1 | - | 9 | 3031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Espnl| Gene Symbol: Espnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033292.3 | NP_001028464.1 | 9 | 3029 | ||
| GenBank mRNA | AK134448.1 | - | 9 | 3031 | ||