siRNA Details
| Gene Symbol: Camsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276359.1 | NP_001263288.1 | 17 | 6331 | ||
| NM_001276360.1 | NP_001263289.1 | 18 | 6364 | |||
| NM_001276361.1 | NP_001263290.1 | 18 | 6391 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Camsap1| Gene Symbol: Camsap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276359.1 | NP_001263288.1 | 17 | 6331 | ||
| NM_001276360.1 | NP_001263289.1 | 18 | 6364 | |||
| NM_001276361.1 | NP_001263290.1 | 18 | 6391 | |||