siRNA Details
| Gene Symbol: Ano3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128103.2 | NP_001121575.1 | 5 | 647 | ||
| GenBank mRNA | AK045930.1 | - | 5 | 648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano3| Gene Symbol: Ano3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128103.2 | NP_001121575.1 | 5 | 647 | ||
| GenBank mRNA | AK045930.1 | - | 5 | 648 | ||