siRNA Details
| Gene Symbol: Bahd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045523.1 | NP_001038988.1 | 7 | 4075 | ||
| GenBank mRNA | AK129243.1 | - | 7 | 4234 | ||
| AK152597.1 | - | 7 | 4014 | |||
| BC047433.1 | - | 6 | 2825 | |||
| BC100358.1 | - | 7 | 4238 | 30848279 | ||

Bahd1
Genomic Map