siRNA Details
| Gene Symbol: Gm561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033297.2 | NP_001028469.1 | 2 | 306 | ||
| GenBank mRNA | AK002610.1 | - | 2 | 307 | ||
| AK010324.1 | - | 2 | 289 | |||
| BC147042.1 | - | 2 | 301 | 9056050 | ||
| BC147043.1 | - | 2 | 301 | 9056078 | ||

Gm561
Genomic Map