siRNA Details
| Gene Symbol: Gm561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033297.2 | NP_001028469.1 | 2 | 412 | ||
| GenBank mRNA | AK002610.1 | - | 2 | 413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm561| Gene Symbol: Gm561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033297.2 | NP_001028469.1 | 2 | 412 | ||
| GenBank mRNA | AK002610.1 | - | 2 | 413 | ||