siRNA Details
| Gene Symbol: Asxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039939.1 | NP_001035028.1 | 13 | 5761 | ||
| GenBank mRNA | AK141208.1 | - | 1 | 2141 | ||
| BC059028.1 | - | 1 | 2373 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asxl1| Gene Symbol: Asxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039939.1 | NP_001035028.1 | 13 | 5761 | ||
| GenBank mRNA | AK141208.1 | - | 1 | 2141 | ||
| BC059028.1 | - | 1 | 2373 | |||