siRNA Details
| Gene Symbol: 9130204L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101461.2 | NP_001094931.1 | 3 | 472 | ||
| GenBank mRNA | AK078932.1 | - | 3 | 473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9130204L05Rik| Gene Symbol: 9130204L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101461.2 | NP_001094931.1 | 3 | 472 | ||
| GenBank mRNA | AK078932.1 | - | 3 | 473 | ||