siRNA Details
| Gene Symbol: Gm4858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034860.2 | NP_001030032.1 | 3 | 1513 | ||
| NM_001177694.1 | NP_001171165.1 | 1 | 1688 | |||
| GenBank mRNA | AK139723.1 | - | 3 | 1514 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4858| Gene Symbol: Gm4858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034860.2 | NP_001030032.1 | 3 | 1513 | ||
| NM_001177694.1 | NP_001171165.1 | 1 | 1688 | |||
| GenBank mRNA | AK139723.1 | - | 3 | 1514 | ||