siRNA Details
Gene Symbol: Gm4858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034860.2 | NP_001030032.1 | 3 | 1063 | ||
NM_001177694.1 | NP_001171165.1 | 1 | 1238 | |||
GenBank mRNA | AK139723.1 | - | 3 | 1064 | ||
BC172161.1 | - | 1 | 1222 |
