siRNA Details
| Gene Symbol: Flg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013804.1 | NP_001013826.1 | 1001 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flg2| Gene Symbol: Flg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013804.1 | NP_001013826.1 | 1001 | |||