siRNA Details
| Gene Symbol: Trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030016.2 | NP_084292.1 | 2 | 1108 | ||
| GenBank mRNA | AK039588.1 | - | 2 | 1109 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trmt13| Gene Symbol: Trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030016.2 | NP_084292.1 | 2 | 1108 | ||
| GenBank mRNA | AK039588.1 | - | 2 | 1109 | ||