siRNA Details
| Gene Symbol: Fam206a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081420.1 | NP_001074889.1 | 6 | 1974 | ||
| GenBank mRNA | AK164596.1 | - | 6 | 1787 | ||
| BC026590.1 | - | 6 | 1953 | 4973099 | ||
| BC062190.1 | - | 7 | 1955 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam206a| Gene Symbol: Fam206a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081420.1 | NP_001074889.1 | 6 | 1974 | ||
| GenBank mRNA | AK164596.1 | - | 6 | 1787 | ||
| BC026590.1 | - | 6 | 1953 | 4973099 | ||
| BC062190.1 | - | 7 | 1955 | |||