siRNA Details
| Gene Symbol: Frrs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142965.1 | NP_001136437.1 | 5 | 1666 | ||
| GenBank mRNA | AK078331.1 | - | 4 | 1329 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frrs1l| Gene Symbol: Frrs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142965.1 | NP_001136437.1 | 5 | 1666 | ||
| GenBank mRNA | AK078331.1 | - | 4 | 1329 | ||