siRNA Details
| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081184.1 | NP_001074653.1 | 44 | 5710 | ||
| GenBank mRNA | AK122556.1 | - | 17 | 5904 | ||
| AK146138.1 | - | 15 | 2092 | |||
| AK146256.1 | - | 15 | 2092 | |||
| BC039622.1 | - | 6 | 1091 | |||
| BC057079.1 | - | 33 | 4188 | |||
| BC089005.1 | - | 42 | 5376 | |||

Focad
Genomic Map