siRNA Details
| Gene Symbol: Zyg11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167936.1 | NP_001161408.1 | 13, 14 | 2101 | ||
| GenBank mRNA | BC022150.1 | - | 11, 12 | 1649 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zyg11a| Gene Symbol: Zyg11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167936.1 | NP_001161408.1 | 13, 14 | 2101 | ||
| GenBank mRNA | BC022150.1 | - | 11, 12 | 1649 | ||