siRNA Details
| Gene Symbol: Zfyve9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183300.2 | NP_899123.2 | 13 | 1638 | ||
| GenBank mRNA | BC038255.1 | - | 2 | 133 | ||
| BC046341.1 | - | 3 | 274 | |||
| BC058416.1 | - | 3 | 198 | |||
| BC079578.1 | - | 3 | 206 | |||
| DQ303413.1 | - | 13 | 1638 | |||

Zfyve9
Genomic Map